missing translation for 'onlineSavingsMsg'
Learn More

C5a Rat anti-Mouse, Unlabeled, Clone: I52-1486, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 558027

Product Code. 15829168

  • ONLINE EXCLUSIVE PRICE
    £156.00
    £149.10 / Each
    Save £6.90
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The purified I52-1486 antibody reacts with the mouse C5a protein. Anaphylatoxin C5a is a bioactive cleavage product released from plasma component C5 during complement activation and is involved in mediation of a variety of cellular immune responses, as well as being potent pro-inflammatory agents. The release of this cleavage product is a reliable indicator of in vivo or in vitro complement activation. Clone I52-1486 hybridoma was generated by the fusion of spleen cells from rats immunized with recombinant mouse C5a protein. The I52-1486 antibody is specific for a neoepitope exposed in mouse C5a/C5adesArg and does not cross-react with mouse C5. This capacity makes antibody I52-1486 a preferential capture antibody for direct detection of mouse C5a from plasma or serum samples.

This antibody is routinely tested by ELISA. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

C5a
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Recombinant mouse C5a
0.1 mg
Immunology
Mouse
IgG1 κ
ELISA
I52-1486
Unconjugated
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.