Learn More
centromere protein M, Mouse, Clone: 4C12-2C8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant C22orf18.
Brand: Abnova H00079019-M01.100ug
Description
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPM is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM
Sequence: MSVLRPLDKLPGLNTATILLVGTEDALLQQLADSMLKEDCASELKVHLAKSLPLPSSVNRPRIDLIVFVVNLHSKYSLQNTEESLRHVDASFFLGKVCFLATGAGRESHCSIHRHTVVKLAHTYQSPLLYCDLEVEGFRATMAQRLVRVLQICAGHVPGVSALNLLSLLRSSEGPSLEDLSpecifications
| centromere protein M | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000705 | |
| CENPM | |
| C22orf18 (AAH00705, 1 a.a. ∼ 180 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 4C12-2C8 | |
| Mouse monoclonal antibody raised against a full length recombinant C22orf18. | |
| CENPM | |
| C22orf18/CENP-M/MGC861/PANE1/bK250D10.2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 79019 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.