missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD4 Rabbit anti-Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540925
This item is not returnable.
View return policy
Description
Peptide sequence: NNGFNSGSGS SSTKLKIQLK RDEENRGSYT EGLHENGVCC SDPLSLLESR Sequence homology: Dog: 86%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 91%; Rat: 93%
C21orf18, located on chromosome 21, is predicted to encode for a hypothetical protein.
Specifications
| SETD4 | |
| Polyclonal | |
| Unconjugated | |
| SETD4 | |
| SETD4 | |
| Synthetic peptide directed towards the C-terminal of human C21ORF18 | |
| 100 μL | |
| Primary | |
| Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NVD3, P58467 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100051888, 100347533, 100726890, 224440, 245975, 487747, 540440, 54093 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction