Learn More
complement component 1, q subcomponent binding protein, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant C1QBP.
Brand: Abnova H00000708-A01.50uL
Description
The human complement subcomponent C1q associates with C1r and C1s in order to yield the first component of the serum complement system. The protein encoded by this gene is known to bind to the globular heads of C1q molecules and inhibit C1 activation. This protein has also been identified as the p32 subunit of pre-mRNA splicing factor SF2, as well as a hyaluronic acid-binding protein. [provided by RefSeq
Sequence: IKNDDGKKALVLDCHYPEDEVGQEDEAESDIFSIREVSFQSTGESEWKDTNYTLNTDSLDWALYDHLMDFLADRGVDNTFADELVELSTALEHQEYITFLEDLKSFVKSQSpecifications
| complement component 1, q subcomponent binding protein | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant C1QBP. | |
| C1QBP | |
| GC1QBP/HABP1/SF2p32/gC1Q-R/gC1qR/p32 | |
| Mouse | |
| 50 μL | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001212 | |
| C1QBP | |
| C1QBP (NP_001203, 173 a.a. ∼ 282 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 708 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.