missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1GALT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543363
This item is not returnable.
View return policy
Description
Peptide sequence: NVEAGDSRDT IGKETFHPFV PEHHLIKGYL PRTFWYWNYN YYPPVEGPGC Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 93%.
Glycosyltransferase that generates the core 1 O-glycan Gal-beta1-3GalNAc-alpha1-Ser/Thr (T antigen), which is a precursor for many extended O-glycans in glycoproteins. Plays a central role in many processes, such as angiogenesis, thrombopoiesis and kidney homeostasis development.
Specifications
| C1GALT1 | |
| Polyclonal | |
| Unconjugated | |
| C1GALT1 | |
| 2210410E06Rik; AV284120; B3Gal-T8; Beta-1,3-galactosyltransferase; C1GALT; C1GALT1; Core 1 beta1,3-galactosyltransferase 1; core 1 beta3-Gal-T; core 1 beta3-Gal-T1; core 1 O-glycan T-synthase; Core 1 synthase; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase (C1galt1); core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 13-galactosyltransferase (C1galt1); glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; Plt1; T-syn; T-synthase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56913 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NS00 | |
| C1GALT1 | |
| Synthetic peptide directed towards the middle region of human C1GALT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction