Learn More
anti-C13ORF18, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00080183-PW1.1Set
Product Code. 16103714
  • £729.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-C13orf18 (300μL), Antibody pair for WB: mouse purified polyclonal anti-C13orf18 (50μg)
Specifications
C13orf18
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
FLJ21562/FLJ43762
Mouse, Rabbit
RUO
80183
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
C13orf18
C13orf18
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-C13orf18 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-C13orf18 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.