missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C18orf14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534724
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C18orf14 gene ontology (GO) annotation includes protein binding.
Specifications
| C18orf14 | |
| Polyclonal | |
| Unconjugated | |
| CCDC102B | |
| ACY1L; AN; c102b; C18orf14; CCDC102B; coiled-coil domain containing 102B; coiled-coil domain-containing protein 102B; HsT1731 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79839 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.97 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q68D86 | |
| CCDC102B | |
| Recombinant protein encompassing a sequence within the center region of human CCDC102B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction