Learn More
c-Jun (pS63) Mouse, Unlabeled, Clone: 2/C-JUN (PS63), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558036
Description
The activator protein transcription factor (AP-1) was identified as a protein that recognizes specific sequences in the cis-control regions of the SV40 virus and the human metallothionein IIA gene. AP-1 is composed of protein products of two different gene families: jun and fos. The AP-1 transcription factor is either a homodimer of Jun proteins or a heterodimer of Jun and Fos proteins. The transcriptional activity of Jun is enhanced by phosphorylation of serines 63 (S63) and 73 in its activation domain. Phosphorylation at both sites is necessary for stimulating the activating function of Jun. Jun is phosphorylated by JNK protein kinases that are activated by the same signals that potentiate Jun activity. The 2/c-Jun/(S63) monoclonal antibody recognizes the phosphorylated S63 of c-Jun.
Host Species: Mouse
Clone: 2
Isotype: IgG1
Species Reactivity: Human
Immunohistochemistry (Formalin-Antigen Retrieval Required), Western Blotting
Specifications
| c-Jun (pS63) | |
| Monoclonal | |
| 0.25mg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Phosphorylated Human c-Jun | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 2/C-JUN (PS63) | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.