missing translation for 'onlineSavingsMsg'
Learn More

c-IAP-2 w/contol Mouse anti-Human, Unlabeled, Clone: F30-2285, BD

Mouse Monoclonal Antibody with Control

£213.00 - £404.00

Specifications

Antigen c-IAP-2 w/contol
Clone F30-2285
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15816858
View Documents
BD Biosciences
552782
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15826858
View Documents
BD Biosciences
552783
150 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Programmed cell death is a normal physiologic process required for maintenance as well as for development in multi-cellular organisms. It is important for apoptosis to be tightly controlled because dysregulation of cell death pathways can lead to pathogenesis. One group of proteins which aids in the regulation of the apoptotic process is called inhibitors of apoptosis (IAPs). This group of proteins acts by directly inhibiting a class of proteins known as the executioners of apoptosis, the caspases. Caspases are inactive cytosolic proteases that upon activation can cause the demise of the cell. IAPs directly inhibit apoptosis by physically interacting with and blocking caspase activity. The first human IAP to be identified, NAIP, was discovered based on its association with a neurodegenerative disorder. Subsequently, six additional human IAPs have been identified, including survivin, XIAP, c-IAP-1, c-IAP-2, BRUCE, and pIAP. These proteins share sequence motifs including a RING zinc finger domain as well two to three copies of an ∼65 amino acid baculovirus IAP repeat (BIR) domain. BIR regions promote protein-protein interaction(s) with caspases and are required for inhibition of caspase activity and apoptosis. While the RING zinc finger regions are not required for this function, they have been found to enhance the caspase inhibitory action of IAPs. Each of these inhibitors displays some specificity with regard to their ability to bind and inhibit caspases. c-IAP-1, c-IAP-2 and XIAP have been shown to block the activity of caspases-3 and -7, while NAIP does not. Thus, IAPs provide a central role in regulation of apoptosis, while subtle differences between the IAPs may confer specificity in the regulation of the various caspases. c-IAP-1 has a molecular weight of ∽72kDa in SDS/PAGE. The antibody recognizes human c-IAP-2. Recombinant human c-IAP-2 expressed in E. coli was used as an immunogen.

Host Species:
Clone: F30-2285
Species Reactivity: Human

Western Blotting

Specifications

Specifications

c-IAP-2 w/contol
Western Blot
Unconjugated
RUO
Human
IgG1
Affinity Purified
F30-2285
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.