Learn More
c-Cbl (pY700) Mouse anti-Human, Unlabeled, Clone: 47/C-CBL, BD
Mouse Monoclonal Antibody
£577.00
Specifications
| Antigen | c-Cbl (pY700) |
|---|---|
| Clone | 47/C-CBL |
| Concentration | 250μg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15876159
|
BD Biosciences
612305 |
150 μg | |||||||
Description
Cbl (Casitas B-lineage lymphoma) was identified in the genome of a transforming retrovirus from a mouse pre-B lymphoma. The cellular gene product c-Cbl is one of numerous Cbl-related proteins found in vertebrate and invertebrate organisms. It is an 120kDa adapter protein that contains multiple functional domains, including a RING finger motif, a tyrosine kinase-binding (TKB) domain, and a proline-rich region. The TKB domain directly interacts with specific auto-phosphorylation sites in activated protein-tyrosine kinases (PTK). Through the RING finger motif, c-Cbl recruits and activates an E2 ubiquitin-conjugating enzyme, thus targeting the activated PTK for protein degradation. The proline-rich region contains SH3 domain-binding and 14-3-3 protein-binding motifs. c-Cbl is also phosphorylated at tyrosines 700 (Y700), 731, and 774 by Syk- and Src-family kinases after the stimulation of some integrins and a wide variety of receptors for antigens, immunoglobulins, growth factors, cytokines, and hormones. In turn, the phosphorylated Y700 site interacts with the SH2 domains of CRK and Vav1. The c-Cbl adapter protein is expressed in the cytoplasm in all tissues, with especially high levels of expression in hematopoietic cells. Through its many functional sites, c-Cbl plays key roles in the positive and negative regulation of vital cell functions, including T Cell Receptor-mediated cellular immune responses.
Immunofluorescence, Western Blotting
Specifications
| c-Cbl (pY700) | |
| 250μg/mL | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG1 | |
| Affinity Purified |
| 47/C-CBL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Human c-Cbl (pY700) Peptide | |
| Primary |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.