missing translation for 'onlineSavingsMsg'
Learn More

Bub3 Mouse, Unlabeled, Clone: 31, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611731

 View more versions of this product

Product Code. 15826019

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Accurate chromosome segregation requires that all pairs of sister chromatids become appropriately attached to mitotic spindles before the onset of anaphase. Cell cycle checkpoints monitor kinetochore-microtubule interactions, so that cell cycle progression can be delayed until proper chromosome attachments are formed. In yeast, Bub1-3 genes are required for proper mitotic delay in response to unattached kinetochores. In mammals, the homologues to yeast Bub1 and Bub3 form a complex that binds kinetochores and has protein kinase activity. Bub3 contains four WD repeats, three in the N-terminus and one in the C-terminus, and a central Bub1-binding domain. During prophase and prometaphase, Bub3 localizes to the kinetochore before attachment to microtubules. In addition, taxol-induced formation of lagging chromosomes due to a delay of cell cycle progression increases the level of Bub3 co-localized with kinetochores, while correctly aligned chromosomes found in metaphase do not exhibit this co-localization. Thus, Bub3, in association with Bub1, may be important for sensing kinetochore attachment to microtubules during the prometaphase to metaphase transition.

Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Bub3 aa. 4-16

Bioimaging, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bub3
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Bub3 aa. 4-16
150 μg
Cell Biology
Human, Mouse, Rat
IgG1
Western Blot
31
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.