missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTF3 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA547771
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes the basic transcription factor 3. This protein forms a stable complex with RNA polymerase IIB and is required for transcriptional initiation. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.
Specifications
| BTF3 | |
| Polyclonal | |
| Unconjugated | |
| BTF3 | |
| 1700054E11Rik; basic transcription factor 3; BETA-NAC; BTF3; BTF3a; BTF3b; NACB; NAC-beta; Nascent polypeptide-associated complex subunit beta; nascent-polypeptide-associated complex beta polypeptide; OK/SW-cl.8; RNA polymerase B transcription factor 3; transcription factor BTF3 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 218490, 294680, 689 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P20290, Q64152 | |
| BTF3 | |
| E. coli-derived recombinant human BTF3 Lys46-Asn206. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction