missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD9 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human BTBD9 protein.
Brand: Abnova H00114781-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MCRALLYGGMRESQPEAEIPLQDTTAEAFTMLLKYIYTGRATLTDEKEEVLLDFLSLAHKYGFPELEDSTSEYLCTILNIQNVCMTFDVASLYSLPKLTCMCCMFMDRNAQEVLSSEGFLSLSKTALLNIVLRDSFAAPEKDIFLALLNWCKHNSKENHAEIMQAVRLPLMSLTELLNVVRPSGLLSPDAILDAIKVRSESRDMDLNYRGMLIPEENIATMKYGAQVVKGELKSALLDGDTQNYDLDHGFSRHPIDDDCRSGIEIKLGQPSIINHIRILLWDRDSRSYSYFIEVSMDELDWVRVIDHSQYLCRSWQKLYFPARVCSGDGVSLWCPLWSRTPELKQSSLLGLPKCRYIRIVGTHNTVNKIFHIVAFECMFTNKTFTLEKGLIVPMENVATIADCASVIEGVSRSRNALLNGDTKNYDWDSGYTCHQLGSGAIVVQLAQPYMIGSIRLLLWDCDDRSYSYYVEVSTNQQQWTMVADRTKVSCKSWQSVTFERQPASFIRIVGTHNTANEVFHCVHFECPEQQSSQKEENSEESGTGDTSLAGQQLDSHALRAPSGSSLPSSPGSNSRSPNRQHQSpecifications
| BTBD9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human BTBD9 protein. | |
| BTBD9 | |
| FLJ32945/KIAA1880/MGC120517/MGC120519/MGC120520/dJ322I12.1 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 114781 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC101357.1 | |
| BTBD9 | |
| BTBD9 (AAI01358.1, 1 a.a. ∼ 582 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction