missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZMYND11 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA527899

Product Code. 13422017

  • £490.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: BS69 tansfected 293T cell. Predicted reactivity: Mouse (97%), Rat (97%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene was first identified by its ability to bind the adenovirus E1A protein. The protein localizes to the nucleus. It functions as a transcriptional repressor, and expression of E1A inhibits this repression. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZMYND11
Polyclonal
Unconjugated
ZMYND11
2210402G22Rik; adenovirus 5 E1A-binding protein; Bone morphogenetic protein receptor-associated molecule 1; BRAM1; BS69; MGC111056; MRD30; Protein BS69; RP11-486H9.1; Unknown (protein for MGC:137432); zinc finger MYND domain-containing protein 11; Zinc finger MYND domain-containing protein 11-like protein; zinc finger MYND-type containing 11; zinc finger, MYND domain containing 11; zinc finger, MYND-type containing 11; ZMYND11
Rabbit
Antigen affinity chromatography
RUO
10771
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q15326
ZMYND11
Recombinant protein encompassing a sequence within the center region of human BS69. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.