missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRMS1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523800
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
The protein encoded by this gene shows sequence similarity to the human breast carcinoma metastasis suppressor (BRMS1) protein and the mammalian Sds3 (suppressor of defective silencing 3) proteins. This protein is a component of the mSin3a family of histone deacetylase complexes (HDAC).
Specifications
| BRMS1L | |
| Polyclonal | |
| BRMS1L | |
| BRMS1, BRMS1-homolog protein p40, BRMS1-like protein p40, breast cancer metastasis-suppressor 1-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 52592, 84312 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Q5PSV4, Q3U1T3 | |
| BRMS1L | |
| KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human BRMS1L | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction