missing translation for 'onlineSavingsMsg'
Learn More

Brm Mouse, Unlabeled, Clone: 24, BD

Mouse Monoclonal Antibody

£268.92

Specifications

Antigen Brm
Clone 24
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15875429
View Documents
BD Biosciences
610390
150 μg
ONLINE EXCLUSIVE PRICE
£280.00
£268.92 / 1 set
Save £11.08
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Several of the SNF and SWI genes of Saccharomyces cerevisiae encode proteins that are involved in the regulation of transcriptional activation. One of these proteins, SNF2/SWI2, has both . The human Brm protein has been reported to be a 180 kDa nuclear factor that acts as a transcriptional activator when fused to a heterologus DNA binding domain. Transfected Brm, expressed in cells lacking endogenous protein, can cooperate with the glucocorticoid receptor (GR) in transcriptional activation. The cooperation between Brm and GR requires the DNA binding domain of GR and the helicase domain and the P/Q-charged domain of Brm. However, Brm does not affect on several other transcription factors. The retinoblastoma protein, Rb, stimulates the transcription of a number of genes. Like Brm, Rb up-regulates glucocorticoid-receptor-mediated transcription. Brm and Rb interact in vitro and in vivo , requiring the Rb-pocket domain and the consensus Rb-binding motif of Brm.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

Brm
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
24
Western Blot
Unconjugated
RUO
Canine, Chicken, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.