Learn More
BrdU (Bromodeoxyuridine) Mouse, Unlabeled, Clone: B44, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 347580
Product Code. 15826108
  • £329.00 / Each
Description
Anti-BrdU, clone B44, is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with spleen cells from BALB/c mice immunized with iodouridine-conjugated ovalbumin.
Bromodeoxyuridine (BrdU) is a uridine derivative that can be incorporated into DNA in place of thymidine. Anti-BrdU identifies BrdU (but not thymidine) in single-stranded DNA, free BrdU, or BrdU coupled to a protein carrier. The antibody also reacts with iodouridine.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BrdU (Bromodeoxyuridine)
Monoclonal
25μg/mL
Phosphate buffered saline with gelatin and 0.1% sodium azide.
Iodouridine
100 Tests
Immunology
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Flow Cytometry
B44
Unconjugated
Mouse
Affinity Purified
RUO
Primary
IgG1 κ