missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541544
This item is not returnable.
View return policy
Description
Peptide sequence: KELAQQVTPG DIVSTYGVRK AMGISIPSPV MENNFVDLTE DTEEPKKTDV Sequence homology: Dog: 86%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Rabbit: 79%; Rat: 86%.
BRD7 acts both as coactivator and as corepressor. For example, it activates the Wnt signaling pathway in a DVL1-dependent manner by negatively regulating the GSK3B phosphotransferase activity. In addition, it is required for TP53-mediated cell-cycle arrest in response to oncogene activation. BRD7 promotes acetylation of TP53 at Lys-382, and thereby promotes efficient recruitment of TP53 to target promoters, inhibiting cell cycle progression from G1 to S phase.
Specifications
| BRD7 | |
| Polyclonal | |
| Unconjugated | |
| BRD7 | |
| 75 kDa bromodomain protein; BP75; BRD7; bromodomain containing 7; bromodomain protein 75 kDa; bromodomain-containing 7; bromodomain-containing 7-like protein; bromodomain-containing protein 7; CELTIX1; NAG4; protein CELTIX-1; protein tyrosine phosphatase, non-receptor type 13 interacting protein; Ptpn13ip | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29117 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NPI1 | |
| BRD7 | |
| Synthetic peptide directed towards the C-terminal of human BRD7 (aa 594-643). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction