missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD4 Mouse anti-Human, Clone: 23476, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 730007
This item is not returnable.
View return policy
Description
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
Specifications
| BRD4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| BRD4 | |
| BRD4-NUT fusion oncoprotein, BRD4-NUT FUSION | |
| Mouse | |
| Purified | |
| RUO | |
| 23476 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| Immunoprecipitation | |
| 23476 | |
| Unconjugated | |
| O60885 | |
| BRD4 | |
| 333-460 of 1362 of BRD4 Protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction