Learn More
Invitrogen™ BOB-1 Monoclonal Antibody (SP92)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA516323
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.
Specifications
| BOB-1 | |
| Monoclonal | |
| Unconjugated | |
| POU2AF1 | |
| B-cell-specific coactivator OBF-1; BOB.1; Bob1; BOB-1; OBF.1; OBF1; Obf-1; OCAB; OCA-B; OCT-binding factor 1; POU class 2 associating factor 1; POU domain class 2-associating factor 1; POU domain, class 2, associating factor 1; Pou2af1 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 5450 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| SP92 | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
| Q16633 | |
| POU2AF1 | |
| Synthetic peptide corresponding to C-terminus of human BOB-1 protein. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.