missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMX Mouse, Unlabeled, Clone: 40, BD
Mouse Monoclonal Antibody
£223.06 - £343.55
Specifications
| Antigen | BMX |
|---|---|
| Clone | 40 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15825649
|
BD Biosciences
610792 |
50 μg |
£520.00 £223.06 / 1 set Save £296.94 57% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15835649
|
BD Biosciences
610793 |
150 μg | |||||||
Description
BMX ( b one m arrow X kinase) is a cytoplasmic tyrosine kinase identified by reverse transcription of mRNA isolated from human bone marrow and mapped to the chromosomal band Xp22.2. The full length protein is 675 amino acids with a tyrosine kinase domain, an amino terminal pleckstrin domain, as well as an SH3 and SH2 domain. Direct comparison of BMX's primary sequence with other kinases showed that this is highly related to the family of BTK/ITK/TEC. BMX kinase is expressed in fetal and adult tissues, with the highest expression in heart, testis, small intestine and colon. It is undetectable in spleen, brain, kidney, and pancreas. Further analysis of mRNA expression showed that BMX is expressed in hematopoietic tissues and neutrophilic granulocytes, and in patients with acute and myeloid leukemia. The levels of BMX mRNA were substantially lower in patients with acute and chronic lymphoid leukemias, thus suggesting that BMX may be important during myelopoiesis.Specifications
| BMX | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 40 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Chicken, Human, Mouse | |
| Human BMX aa. 138-276 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title