Learn More
Faster™ Anti-blowback Valve
Product Code. 15513376
Click to view available options
For Use With (Equipment):
Faster Safety Cabinet
Product Code. 15513376
Brand: Faster™ FX0002124800
Anti-blowback valve for the Faster™ Safety Cabinet range.
  • Valve is required for safety cabinets that are ducted
  • Prevents external air entering the cabinet back down the ducting so that is doesn't compromise sterility
Specifications
For Use With (Application) Ducting
For Use With (Equipment) Faster Safety Cabinet