Learn More
anti-beta Catenin , Polyclonal, Abcam™
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Chicken, Human, Pig, Xenopus laevis, Zebrafish, Botryllus schlosseri
Immunogen: Synthetic peptide: PGDSNQLAWFDTDL conjugated to KLH, corresponding to amino acids 768-781 of Human or mouse β Catenin.
Applications:
Dot Blotting, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | beta Catenin |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide: PGDSNQLAWFDTDL conjugated to KLH, corresponding to amino acids 768-781 of Human or mouse β Catenin. |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Target Species | B. schlosseri, Chicken, Human, Mouse, Pig, Rat, Xenopus species, Zebrafish |
| Form | Antisera |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.