missing translation for 'onlineSavingsMsg'
Learn More

beta Actin Loading Control Monoclonal Antibody (BA3R), DyLight™ 488
GREENER_CHOICE

Product Code. 15577231
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15577231 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577231 Supplier Invitrogen Supplier No. MA515739D488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

beta Actin Loading Control Monoclonal Antibody (BA3R), DyLight™ 488

beta Actin Loading Control Monoclonal Antibody for Western Blot, ICC/IF

MA5-15739-D488 detects beta actin from human, mouse, rat, rabbit, and chicken samples. MA5-15739-D488 has been successfully used in immunofluorescence. DyLight 488 has an excitation/emission of 493/518 nm. For visualization of actin filaments by immunofluorescence, fixation and permeabilization with methanol is required.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta Actin Loading Control
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone BA3R
Concentration 1mg/mL
Conjugate DyLight 488
Formulation PBS with no preservative
Gene ACTB
Gene Accession No. P60706, P60709, P60710, P29751, P60711
Gene Alias Actin, ACTB, Atcb
Gene Symbols ACTB
Host Species Mouse
Immunogen Beta-actin N-terminal peptide.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009272, 11461, 396526, 60, 81822
Target Species Chicken, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4° C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.