Learn More
anti-beta Actin, Clone: mAbcam 8226, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB8226-100ug
Description
Host Species: Mouse
Clone: mAbcam 8226
Isotype: IgG1, κ
Cross Reactivity: Mouse, Rat, Rabbit, Chicken, Cow, Dog, Human, Pig, African Green Monkey, Chinese Hamster, Armenian Hamster. Predicted to work with Sheep, Horse, Guinea pig
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human beta Actin
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
| beta Actin | |
| mAbcam 8226 | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human beta Actin | |
| Isotype Loading Control, Signal Transduction, Tags And Cell Markers | |
| Bovine, Canine, Chicken, Equine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep |
| Monoclonal | |
| 0.5mg/mL | |
| Mouse | |
| 100 μL | |
| Primary | |
| IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.