Learn More
anti-beta Actin, Clone: mAbcam 8224, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB8224-100UG
Description
Host Species: Mouse
Clone: mAbcam 8224
Isotype: IgG3, κ
Cross Reactivity: Mouse, Rat, Rabbit, Chicken, Cow, Cat, Dog, Human, Pig, Saccharomyces cerevisiae, Xenopus laevis, Fruit fly (Drosophila melanogaster), Schizosaccharomyces pombe, Chinese Hamster, Other
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human beta Actin.
Applications:
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
Specifications
| beta Actin | |
| mAbcam 8224 | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human beta Actin. | |
| Isotype Loading Control, Signal Transduction, Tags And Cell Markers | |
| Bovine, Canine, Chicken, Drosophila, Feline, Hamster, Human, Mouse, Pig, Rabbit, Rat, S. cerevisiae, S. pombe, Xenopus species |
| Monoclonal | |
| 0.6mg/mL | |
| Mouse | |
| 100 μg | |
| Primary | |
| IgG3 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.