Learn More
Bcl-xL Mouse anti-Human, Mouse, Clone: 7D9, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA137166
Description
This antibody is predicted to react with rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is Hodgkin's lymphoma.
BCL-XL is a part of the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. BCL-XL is located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential and controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. The longer isoform acts as an apoptotic inhibitor and the shorter form acts as an apoptotic activator.
Specifications
| BCL-XL | |
| Monoclonal | |
| Unconjugated | |
| Q07817, Q64373 | |
| BCL2L1 | |
| Synthetic peptide corresponding to N-terminus of mouse BCL-XL protein | |
| 1 mL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 7D9 | |
| BCL2L1 | |
| BCL-XL/S, BCL2L, BCLX, Bcl-X, DKFZp781P2092, bcl-xL, bcl-xS | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 12048, 598 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.