missing translation for 'onlineSavingsMsg'
Learn More

Bcl-X Mouse, Unlabeled, Clone: 44, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610746

 View more versions of this product

Product Code. 15865619

  • ONLINE EXCLUSIVE PRICE
    £233.00
    £223.06 / 1 set
    Save £9.94
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Bcl-x is related to the Bcl-2 protein and can function independently of Bcl-2 in regulating apoptosis (programmed cell death). The bcl-x transcript is expressed in a number of tissues, with the highest levels in the lymphoid and central nervous systems. Two distinct cDNA species, bcl-xL and bcl-xS, have been observed and appear to arise from alternate 5' splice sites located within the first coding exon of the bcl-x gene. Bcl-xL is composed of 233 amino acids and is similar in size and structure to Bcl-2. The Bcl-xS polypeptide shows a deletion of 63 amino acids which consist of the region of Bcl-xL with the highest degree of amino acid identity to Bcl-2. Like Bcl-2, Bcl-xL inhibits cell death upon growth factor withdrawal when transfected into an IL-3-dependent cell line. However, Bcl-xS inhibits Bcl-2 mediated cell survival.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bcl-X
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Bcl-Xl aa. 18-233
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Western Blot
44
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.