Learn More
Bcl-2 Hamster anti-Human, FITC, Clone: 6C8, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 554234
Includes: FITC Hamster Anti-Human Bcl-2 and FITC Hamster Isotype Control (Anti-KLH)
Description
Programmed cell death (apoptosis) is a normal physiologic process which occurs during embryonic development as well as in maintenance of tissue homeostasis. The apoptotic program is characterized by certain morphological features. These include changes in the plasma membrane such as loss of membrane asymmetry and attachment, a condensation of the cytoplasm and nucleus, and internucleosomal cleavage of DNA. In the final stages, the dying cells become fragmented into “apoptotic bodies” which are rapidly eliminated by phagocytic cells without eliciting significant inflammatory damage to surrounding cells. Members of the Bcl-2 family play a major role in regulating the cellular response to apoptotic signals. Bcl-2 is considered to be a novel proto-oncogene because it blocks apoptosis in many cell types. Bcl-2 is thought to provide selective survival advantage for cells by blocking apoptosis and thus may contribute to tumorigenesis. Bcl-2 is a 26 kDa intracellular, integral membrane protein found primarily in the nuclear envelope, endoplasmic reticulum and outer mitochondrial membrane.
Specifications
| Bcl-2 | |
| Monoclonal | |
| FITC | |
| Human Bcl-2 Recombinant Protein, Keyhole limpet hemocyanin | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgG |
| Flow Cytometry | |
| 6C8 | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.