Learn More
Bcl-2 Set Hamster anti-Mouse, PE, Clone: 3F11, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 556537
Description
Programmed cell death (apoptosis) is a normal physiologic process which occurs during embryonic development as well as in maintenence of tissue homeostasis. The apoptotic program is characterized by certain morphological features. These include changes in the plasma membrane such as loss of membrane asymmetry and attachment, a condensation of the cytoplasm and nucleus, and internucleosomal cleavage of DNA. In the final stages, the dying cells become fragmented into “apoptotic bodies” which are rapidly eliminated by phagocytic cells without eliciting significant inflammatory damage to surrounding cells. Members of the Bcl-2 family play a major role in regulating the cellular response to apoptotic signals. Bcl-2 is considered to be a novel proto-oncogene because it blocks apoptosis in many cell types. Bcl-2 is thought to provide selective survival advantage for cells by blocking apoptosis and thus may contribute to tumorigenesis. Bcl-2 is a 26 kDa intracellular, integral membrane protein found primarily in the nuclear envelope, endoplasmic reticulum and outer mitochondrial membrane.
Clone 3F11 reacts with mouse Bcl-2. It does not cross-react with human Bcl-2. Gel purified recombinant mouse Bcl-2 protein was used as immunogen.
Clone A19-3 is an Armenian hamster IgG isotype control which is specific for the hapten trinitrophenol (TNP). This hapten is not expressed on human cells or human cell lines.
Specifications
| Bcl-2 Set | |
| Monoclonal | |
| PE | |
| Affinity Purified | |
| RUO | |
| PE Hamster Anti-Mouse Bcl-2 (100 tests), PE Hamster IgG Isotype Control (Anti-Trinitrophenol (TNP)) (100 tests) | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 3F11 | |
| Hamster | |
| 100 Tests | |
| Primary | |
| Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.