Learn More
Bcl-2 Mouse, Unlabeled, Clone: 7, BD
Mouse Monoclonal Antibody
£223.06 - £314.49
Specifications
| Antigen | Bcl-2 |
|---|---|
| Clone | 7 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15845509
|
BD Biosciences
610538 |
50 μg |
£233.00 £223.06 / 1 set Save £9.94 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15855509
|
BD Biosciences
610539 |
150 μg |
£328.00 £314.49 / 1 set Save £13.51 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human hematopoietic malignancies are often characterized by gene rearrangements which deregulate gene expression. The Bcl-2 gene (B-cell leukemia/lymphoma 2) was discovered by walking the chromosomal breakpoints t(14,18). This gene consists of two exons and several transcripts that encode two different protein products, Bcl-2α and Bcl-2β. Both are approximately 26-30kDa and differ at their C-terminal region. Bcl-2 suppresses programmed cell death (apoptosis) and this property appears to be modulated by homodimerization or association with Bax and Bad, two cell death promoters.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Bcl-2 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 7 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.