Learn More
Bax Mouse, Unlabeled, Clone: 6A7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556467
Description
The Bcl-2 family members are involved in mediating programmed cell death or apoptosis, and share two highly conserved functional regions, Bcl-2 homology 1 and 2 (BH1 and BH2). Several of the family members including Bcl-2 act as inhibitors of apoptosis, whereas others such as Bax promote cell death. It is thought that protein-protein interactions between Bcl-2 family members play an important role in their function. For example, Bax may form homodimers or heterodimers with either Bcl-2 and Bcl-XL (long). Bax homodimers is thought to promote cell death, whereas Bax heterodimerization with either Bcl-2 and Bcl-XL appears to block cell death. When Bax is present in excess, it can counteract the ability of Bcl-2 to inhibit cell death. Bax expression has been identified in a variety of tissues, including lung, stomach, kidney, thymus, lymph nodes, bone marrow, spleen, heart, exocrine pancreas, and brain. Liver, kidney, and the exocrine pancreas have been found to contain little or no Bcl-2. The Bax gene potentially encodes three different proteins Bax-α (21 kDa), Bax-β (24 kDa) and Bax-γ (5 kDa). Bax-α is the most common transcript.
Specifications
| Bax | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| N-terminal peptide sequence | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 κ |
| Western Blot | |
| 6A7 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.