Learn More
anti-BATF, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00010538-PW1.1Set
Product Code. 16042419
  • £729.00 / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-BATF (300μg), Antibody pair for WB: rabbit polyclonal anti-BATF (50μL)
Specifications
BATF
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
B-ATF/BATF1/SFA-2/SFA2
Mouse, Rabbit
RUO
10538
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
BATF
BATF
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-BATF (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-BATF (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.