missing translation for 'onlineSavingsMsg'
Learn More

basic leucine zipper transcription factor, ATF-like, Mouse, Clone: 1G4, Abnova™

Mouse monoclonal antibody raised against a partial recombinant BATF.

Brand:  Abnova H00010538-M03.100ug

Product Code. 16102746

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events. [provided by RefSeq

Sequence: EKNRIAAQKSRQRQTQKADTLHLESEDLEKQNAALRKEIKQLTEELKYFTSVLNSHEPLCSVLAASTPSPPEVVYSAHAFHQPHVSSPRFQP
Specifications

Specifications

basic leucine zipper transcription factor, ATF-like
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_006399
BATF
BATF (NP_006390, 34 a.a. ∼ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Transcription Regulation
Primary
Human, Mouse, Rat
Antibody
ELISA, Western Blot
1G4
Mouse monoclonal antibody raised against a partial recombinant BATF.
BATF
B-ATF/BATF1/SFA-2/SFA2
Mouse
Affinity Purified
RUO
Yes
10538
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.