missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAT5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531342
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (98%), Rat (97%), Pig (98%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is thought to be involved in some aspects of immunity.
Specifications
| BAT5 | |
| Polyclonal | |
| Unconjugated | |
| ABHD16A | |
| Abhd16a; abhydrolase domain containing 16A; abhydrolase domain-containing protein 16A; AI326074; Alpha/beta hydrolase domain-containing protein 16A; Bat5; Bat-5; D17H6S82E; D6S82E; G5; hBAT5; HLA-B associated transcript 5; HLA-B-associated transcript 5; HLA-B-associated transcript 5 homolog; mBAT5; Monoacylglycerol lipase ABHD16A; Ng26; Phosphatidylserine lipase ABHD16A; PP199; protein ABHD16A; protein BAT5; protein G5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.01 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95870 | |
| ABHD16A | |
| Recombinant protein encompassing a sequence within the center region of human BAT5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction