Learn More
BARX homeobox 1, Mouse, Clone: 1E7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant BARX1.
Brand: Abnova H00056033-M01.100ug
Description
This gene encodes a member of the Bar subclass of homeobox transcription factors. Studies of the mouse and chick homolog suggest the encoded protein may play a role in developing teeth and craniofacial mesenchyme of neural crest origin. The protein may also be associated with differentiation of stomach epithelia. [provided by RefSeq
Sequence: MGLEKRFEKQKYLSTPDRIDLAESLGLSQLQVKTWYQNRRMKWKKIVLQGGGLESPTKPKGRPKKNSIPTSEQLTEQERAKDAEKPAEVPGEPSDRSREDSpecifications
| BARX homeobox 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC009458 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 56033 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1E7 | |
| Mouse monoclonal antibody raised against a full length recombinant BARX1. | |
| BARX1 | |
| BARX1 | |
| BARX1 (AAH09458, 1 a.a. ∼ 100 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.