missing translation for 'onlineSavingsMsg'
Learn More

BAI2, Mouse, Clone: 6A12, Abnova™

Mouse monoclonal antibody raised against a partial recombinant BAI2.

Brand:  Abnova H00000576-M01.100ug

Product Code. 16175144

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis. [provided by RefSeq

Sequence: DPAPSACSALASGVLYGAFSLQDLFPTIASGCSWTLENPDPTKYSLYLRFNRQEQVCAHFAPRLLPLDHYLVNFTCLRPSPEEAVAQ
Specifications

Specifications

BAI2
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_001703
Mouse
Affinity chromatography
RUO
576
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
ELISA, Western Blot
6A12
Mouse monoclonal antibody raised against a partial recombinant BAI2.
BAI2
BAI2
BAI2 (NP_001694, 22 a.a. ∼ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.