missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Anti-bacterial, Anti-odor Insoles

Product Code. 15342800
Change view
Click to view available options
Size:
T38
T43
T44
Unit Size:
10 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15342800 T43 10 pairs
15342790 T38 10 pairs
15352800 T44 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15342800 Supplier Honeywell™ Supplier No. 600031043/7
Quantity Price
1 £66.56 / 10 pairs
10 £61.43 / 10 pairs

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Double-action, perforated blue insoles prevent bacteria and odor.

  • Dual-action: prevents bacteria and odor
  • Perforated for breathability
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With (Application) Insoles for comfort, prevent bacteria/odor
Size T43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.