missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Anti-bacterial, Anti-odor Insoles
Click to view available options
Size:
T38
T43
T44
Unit Size:
10 pairs
Description
- Dual-action: prevents bacteria and odor
- Perforated for breathability
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Insoles for comfort, prevent bacteria/odor |
| Size | T38 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction