missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD anti-Avidin-Fluorescein Isothiocyanate, FITC, Clone: Avidin
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
Description
Flow Cytometry
Specifications
Specifications
| Antigen | Avidin |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC (Fluorescein) |
| Quantity | 0.5 mg |
| Regulatory Status | For research use only (RUO) |
| Storage Buffer | Aqueous buffered solution containing less than or equal to 0.09% sodium azide |
| Content And Storage | Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |
| For Use With (Application) | Flow Cytometry |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction