missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENPP2 Polyclonal Antibody
Description
A suggested positive control for this product is human brain lysate.
The protein encoded by this gene functions as both a phosphodiesterase, which cleaves phosphodiester bonds at the 5' end of oligonucleotides, and a phospholipase, which catalyzes production of lysophosphatidic acid (LPA) in extracellular fluids. LPA evokes growth factor-like responses including stimulation of cell proliferation and chemotaxis. This gene product stimulates the motility of tumor cells and has angiogenic properties, and its expression is upregulated in several kinds of carcinomas. The gene product is secreted and further processed to make the biologically active form. Several alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
| Antigen | ENPP2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | ENPP2 |
| Gene Accession No. | Q13822 |
| Gene Alias | ATX; ATX-X; autotaxin; AutotaxinATX; autotaxin-t; ectonucleotide pyrophosphatase/phosphodiesterase 2; ectonucleotide pyrophosphatase/phosphodiesterase family member 2; E-NPP 2; Enpp2; Extracellular lysophospholipase D; FLJ26803; lysoPLD; NPP2; Npps2; PD-IALPHA; PDNP2; phosphodiesterase I/nucleotide pyrophosphatase 2; phosphodiesterase I/nucleotide pyrophosphatase 2 (autotaxin); plasma lysophospholipase D |
| Gene Symbols | ENPP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?