Learn More
Aurora A Mouse anti-Human, Mouse, Clone: 35C1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180399
Description
Clone 35C1 specifically recognizes an epitope within the non-catalytic N-terminal domain of Aurora-A, and does not inhibit Aurora-A kinase activity. A suggested positive control for immunohistochemical applications is human 293 and mouse llc1 cell lines. In Western blot, this antibody detects a band at ∽46kDa in human HeLa and mouse M-ICc12 cell lysates.
Aurora A is a 46 kDa protein that plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment. Tissue specificity: Highly expressed in testis and weakly in skeletal muscle, thymus and spleen. Also highly expressed in colon, ovarian, prostate, neuroblastoma, breast and cervical cancer cell lines. Multiple transcript variants encoding the same protein have been found for this gene. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal.
Specifications
| Aurora A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O14965, P97477 | |
| AURKA | |
| Recombit Aurora A | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| ChIP Assay, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 35C1 | |
| Unconjugated | |
| AURKA | |
| AIK, ARK1, AURA, AURORA2, BTAK, MGC34538, STK15, STK6, STK7 | |
| Mouse | |
| Protein G | |
| RUO | |
| 20878, 6790 | |
| 4°C or -20°C if preferred | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.