missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPase, H+ transporting, lysosomal, V0 subunit d2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ATP6V0D2.
Brand: Abnova H00245972-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: ETLYPTFGKLYPEGLRLLAQAEDFDQMKNVADHYGVYKPLFEAVGGSGGKTLEDVFYEREVQMNVLAFNSpecifications
| ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ATP6V0D2. | |
| ATP6V0D2 | |
| ATP6D2/FLJ38708/VMA6 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 245972 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_152565 | |
| ATP6V0D2 | |
| ATP6V0D2 (NP_689778, 238 a.a. ∼ 306 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction