Learn More
ATP2A1, Mouse, Clone: 3H4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ATP2A1.
Brand: Abnova H00000487-M06A.200uL
Description
This gene encodes one of the SERCA Ca(2+)-ATPases, which are intracellular pumps located in the sarcoplasmic or endoplasmic reticula of muscle cells. This enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen, and is involved in muscular excitation and contraction. Mutations in this gene cause some autosomal recessive forms of Brody disease, characterized by increasing impairment of muscular relaxation during exercise. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: IDRCNYVRVGTTRVPLTGPVKEKIMAVIKEWGTGRDTLRCLALATRDTPPKREEMVLDDSARFLEYETDLTFVGVVGMLDPPRKEVTGSIQSpecifications
| ATP2A1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_173201 | |
| ATP2A1 | |
| ATP2A1 (NP_775293, 522 a.a. ∼ 612 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 487 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA | |
| 3H4 | |
| Mouse monoclonal antibody raised against a partial recombinant ATP2A1. | |
| ATP2A1 | |
| ATP2A/SERCA1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.