Learn More
ATP13A2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ATP13A2.
Brand: Abnova H00023400-A01.50uL
Description
This gene encodes a member of the P5 subfamily of ATPases which transports inorganic cations as well as other substrates. Mutations in this gene are associated with Kufor-Rakeb syndrome (KRS), also referred to as Parkinson disease 9. Multiple transcript variants encoding different isoforms have been found for this gene
Sequence: KPLWGVRLRLRPCNLAHAETLVIEIRDKEDSSWQLFTVQVQTEAIGEGSLEPSPQSQAEDGRSQAAVGAVPEGAWKDTAQLHKSEEASpecifications
| ATP13A2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ATP13A2. | |
| ATP13A2 | |
| FLJ26510/HSA9947/KRPPD/PARK9 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_022089 | |
| ATP13A2 | |
| ATP13A2 (NP_071372, 68 a.a. ∼ 154 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 23400 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.