missing translation for 'onlineSavingsMsg'
Learn More

ATP Synthase β Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612518

 View more versions of this product

Product Code. 15886199

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

ATP synthase is a large enzyme complex that uses an electrochemical H+ or Na+ gradient to synthesize ATP from ADP and Pi, providing the organism with the ATP needed for energy. The complex consists of two major units, F0 and F1. F0 is embedded in the inner membrane of the mitochondria and, due to its hydrophobic nature, translocates protons across this membrane. F1 is the catalytic portion in the matrix region of the mitochondria and is comprised of α, β, γ, δ, and ε subunits at a 3:3:1:1:1 ratio. The β subunit is synthesized in the nuclear genome,transported to the mitochondria, and assembled with the other subunits. It is encoded by a single copy gene, is ubiquitously expressed and highly conserved among species. ATP synthase β contains an Ets domain binding site, which is a main site for promoter activity. Ets proteins contain domains that are involved in transcriptional activation, protein-protein interactions, and intramolecular repression of DNA binding. This site acts as a target for transcriptional control by the Ets family of transcription factors. Thus, ATP synthase β is involved in the synthesis of ATP and is controlled in part by ETS family proteins.

Bioimaging, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP Synthase β
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human ATP Synthase β aa. 428-539
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
10
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.