Learn More
ATOX1, Mouse, Clone: 2E6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ATOX1.
Brand: Abnova H00000475-M01.100ug
Description
This gene encodes a copper chaperone that plays a role in copper homeostasis by binding and transporting cytosolic copper to ATPase proteins in the trans-Golgi network for later incorporation to the ceruloplasmin. This protein also functions as an antioxidant against superoxide and hydrogen peroxide, and therefore, may play a significant role in cancer carcinogenesis. Because of its cytogenetic location, this gene represents a candidate gene for 5q-syndrome. [provided by RefSeq
Sequence: MPKHEFSVDMTCGGCAEAVSRVLNKLGGVKYDIDLPNKKVCIESEHSMDTLLATLKKTGKTVSYLGLESpecifications
| ATOX1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004045 | |
| ATOX1 | |
| ATOX1 (NP_004036, 1 a.a. ∼ 68 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 2E6 | |
| Mouse monoclonal antibody raised against a partial recombinant ATOX1. | |
| ATOX1 | |
| ATX1/HAH1/MGC138453/MGC138455 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 475 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.