missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535204
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with chicken, mouse and rat based on sequence homology.
This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vacuolar membranes. The protein shows homology to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.Specifications
ATG7 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O95352 | |
ATG7 | |
KLH conjugated synthetic peptide between 540-569 amino acids from the C-terminal region of human ATG7 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ATG7 | |
1810013K23Rik; Agp7; APG7L; APG7-LIKE; ATG12-activating enzyme E1 ATG7; Atg7; ATG7 autophagy related 7 homolog; Atg7l; autophagy related 7; autophagy related 7 homolog; autophagy-related 7; autophagy-related protein 7; DKFZp434N0735; Gm21553; GSA7; hAGP7; ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10533 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |