Learn More
Invitrogen™ Ataxin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522369
Description
Recommended positive controls: SK-N-SH, SK-N-AS, mouse brain, human Ataxin3-transfected 293T. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (85%), Pig (99%), Chicken (89%), Sheep (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Machado-Joseph disease is an autosomal dominant neurologic disorder, and is now known to be the same as previously described spinocerebellar ataxia-3. MJD protein (Ataxin-3) contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is a negative correlation between the age of onset and CAG repeat numbers. This protein interacts with key regulators (CBP, p300 and PCAF) of transcription and represses transcription, and also acts as a histone-binding protein that regulates transcription. MJD is a deubiquitinating enzyme.
Specifications
| Ataxin 3 | |
| Polyclonal | |
| Unconjugated | |
| ATXN3 | |
| 2210008M02Rik; AI463012; AI647473; AT3; ataxin 3; ataxin 3 variant an; ataxin 3 variant ao; ataxin 3 variant at; ataxin 3 variant e; ataxin 3 variant h; ataxin 3 variant m; ataxin 3 variant r; ataxin 3 variant ref; ataxin 3 variant y; ataxin3; ataxin-3; ATX3; ATXN3; JOS; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3); Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1; Machado-Joseph disease protein 1 homolog; MJD; MJD1; olivopontocerebellar ataxia 3; Rsca3; Sca3; spin; Spinocerebellar ataxia type 3 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110616, 4287 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.36 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P54252, Q9CVD2 | |
| ATXN3 | |
| Recombinant protein encompassing a sequence within the center region of human Ataxin 3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.