missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Astrovirus Type 2 Monoclonal Antibody (8E7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516293
This item is not returnable.
View return policy
Description
Astrovirus Type 2 Monoclonal Antibody for ELISA, LF
Astroviruses have a star-like appearance with five or six points and are non-enveloped, non-segmented RNA viruses (single stranded, positive sense) contained within an icosahedral capsid. The genome is relatively small and has three open reading frames (ORFs). There is no vaccine or anti-viral treatment against the Astrovirus infection and has a very high mutation rate. Human astroviruses have been shown in numerous studies to be an important cause of gastroenteritis in young children worldwide.
Specifications
| Astrovirus Type 2 | |
| Monoclonal | |
| 7.44 mg/mL | |
| PBS with 15mM sodium azide; pH 7.5 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Lateral Flow | |
| 8E7 | |
| Unconjugated | |
| Astro virus; Astroviridae; Astrovirus 2 | |
| Astrovirus group 2 grown in LLC MK2 cells. | |
| 2 mg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction